3-Chloroisatoic anhydride 95+%

Supplier: Ambeed
Warning

Synonyms: 8-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione

A829572-1G A829572-5G
77036-574EA 39.26 USD
77036-574 77036-576
3-Chloroisatoic anhydride 95+%
3-Chloroisatoic anhydride

Precaución: for Research Use Only

Formula: C₈H₄ClNO₃
MW: 197.5753 g/mol
MDL Number: MFCD06809785
CAS Number: 63497-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR