H-Dap(Boc)-OH 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Boc-amino)-L-alanine

A829009-100MG A829009-1G A829009-5G A829009-250MG A829009-25G
76993-476EA 8.49 USD
76993-476 76993-478 76993-480 77684-783 77684-784
H-Dap(Boc)-OH 98%
H-Dap(Boc)-OH

Precaución: For research use only.

Formula: C₈H₁₆N₂O₄
MW: 204.2 g/mol
MDL Number: MFCD02683451
CAS Number: 74536-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR