4-(4-Fluorophenoxy)-2-methylaniline 97%

Supplier: Ambeed
Danger

A828026-1G A828026-250MG
77006-736EA 240 USD
77006-736 77882-752
4-(4-Fluorophenoxy)-2-methylaniline 97%
4-(4-Fluorophenoxy)-2-methylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₂FNO
MDL Number: MFCD08687896
CAS Number: 946786-20-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR