1-Benzyl-1H-pyrazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A827686-250MG A827686-5G A827686-100MG A827686-1G
77036-570EA 35 USD
77036-570 77036-572 77674-226 77873-294
1-Benzyl-1H-pyrazole-4-carboxylic acid 98%
1-Benzyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD05864591
CAS Number: 401647-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR