3,5-Dibromo-2-fluoro-6-methylpyridine 97+%
Supplier: Ambeed
|
Warning
|
A827563-100MG
A827563-250MG
77006-728EA
5
USD
77006-728
77006-730
77006-732
77006-734
3,5-Dibromo-2-fluoro-6-methylpyridine 97+%
3,5-Dibromo-2-fluoro-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₄NBr₂F |
MDL Number:
MFCD09475419 CAS Number: 632628-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...