7-Chloro-3-methylbenzo[b]thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A827429-100MG A827429-1G A827429-250MG
77019-558EA 107.16 USD
77019-558 77019-560 77019-562
7-Chloro-3-methylbenzo[b]thiophene-2-carboxylic acid 98%
7-Chloro-3-methylbenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇ClO₂S
MW: 226.67 g/mol
MDL Number: MFCD11104814
CAS Number: 752135-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR