(2-Fluoro-4-(trifluoromethyl)phenyl)hydrazine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A826689-100MG
A826689-250MG
A826689-1G
77036-566EA
46
USD
77036-566
77036-568
77670-180
(2-Fluoro-4-(trifluoromethyl)phenyl)hydrazine hydrochloride 95%
(2-Fluoro-4-(trifluoromethyl)phenyl)hydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₇ClF₄N₂ MW: 230.59 g/mol |
MDL Number:
MFCD03094208 CAS Number: 1030313-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...