N-Ethylbenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: N-Benzylethylamine, N-Benzylethanamine

A826336-100G A826336-25G A826336-500G A826336-5G
77006-720EA 26.53 USD
77006-720 77006-722 77006-724 77006-726
N-Ethylbenzylamine 98%
N-Ethylbenzylamine

Precaución: for Research Use Only

Formula: C₆H₅CH₂NHCH₂CH₃
MW: 135.21 g/mol
MDL Number: MFCD00009031
CAS Number: 14321-27-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR