Methyl-4-trifluoromethylbenzoylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionoate, 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionic acid methyll ester, Methyl 4-(Trifluoromethyl)benzoylacetate, 4-(Trifluoromethyl)benzoylacetic acid methyll ester, Methyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate

A825543-100G A825543-25G A825543-5G
77006-714EA 449.83 USD
77006-714 77006-716 77006-718
Methyl-4-trifluoromethylbenzoylacetate 98%
Methyl-4-trifluoromethylbenzoylacetate

Precaución: for Research Use Only

Formula: C₁₁H₉F₃O₃
MW: 246.185 g/mol
MDL Number: MFCD00216521
CAS Number: 212755-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR