N-Fmoc-O-benzyl-D-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Tyr(Bzl)-OH

A825512-25G A825512-5G A825512-1G
77006-710EA 160 USD
77006-710 77006-712 77882-749
N-Fmoc-O-benzyl-D-tyrosine 98%
N-Fmoc-O-benzyl-D-tyrosine

Precaución: for Research Use Only

Formula: C₃₁H₂₇NO₅
MDL Number: MFCD00235837
CAS Number: 138775-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR