3-Iodo-2-methyl-2H-indazole 95+%

Supplier: Ambeed
Warning

A824368-100MG A824368-1G A824368-250MG A824368-5G
77036-556EA 31.83 USD
77036-556 77036-558 77036-560 77036-562
3-Iodo-2-methyl-2H-indazole 95+%
3-Iodo-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇IN₂
MW: 258.06 g/mol
MDL Number: MFCD12828021
CAS Number: 49572-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR