3-Methylpicolinic acid hydrochloride 98%

Supplier: Ambeed
Warning

A823701-1G A823701-5G
77006-698EA 16.98 USD
77006-698 77006-700
3-Methylpicolinic acid hydrochloride 98%
3-Methylpicolinic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.59 g/mol
MDL Number: MFCD04114110
CAS Number: 123811-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR