(R)-3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A823668-100MG A823668-250MG
77019-540EA 105.03 USD
77019-540 77019-542
(R)-3-(4-Fluorophenoxy)pyrrolidine hydrochloride 95%
(R)-3-(4-Fluorophenoxy)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClFNO
MW: 217.66 g/mol
MDL Number: MFCD11849812
CAS Number: 1314419-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR