1-Aminomethyl-1-(boc-amino)cyclopropane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (1-(aminomethyl)cyclopropyl)carbamate
A823611-100MG
A823611-250MG
A823611-1G
A823611-5G
76993-450EA
60.48
USD
76993-450
76993-452
77573-630
77684-781
1-Aminomethyl-1-(boc-amino)cyclopropane 98%
1-Aminomethyl-1-(boc-amino)cyclopropane
Precaución: For research use only.
Formula:
C₉H₁₈N₂O₂ MW: 186.255 g/mol |
MDL Number:
MFCD12404931 CAS Number: 1027338-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...