1-(3-Bromopropyl)-4-methoxybenzene 95%

Supplier: Ambeed
Danger

A822151-1G A822151-250MG A822151-5G
77036-548EA 152.77 USD
77036-548 77036-550 77036-552
1-(3-Bromopropyl)-4-methoxybenzene 95%
1-(3-Bromopropyl)-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
MDL Number: MFCD00155066
CAS Number: 57293-19-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR