ABT-737 98%

Supplier: Ambeed
Warning

Synonyms: ABT737, (R)-4-(4-((4'-Chloro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((4-((4-(dimethylamino)-1-(phenylthio)butan-2-yl)amino)-3-nitrophenyl)sulphonyl)benzamide

A821402-100MG A821402-1MG A821402-5MG A821402-10MG
77036-542EA 695 USD
77036-542 77036-544 77036-546 77670-164
ABT-737 98%
ABT-737

Precaución: for Research Use Only

Formula: C₄₂H₄₅ClN₆O₅S₂
MW: 813.4 g/mol
MDL Number: MFCD12756212
CAS Number: 852808-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR