4-(Allylamino)benzoic acid 95%

Supplier: Ambeed
Warning

A821218-100MG A821218-1G
77019-524EA 56 USD
77019-524 77019-526
4-(Allylamino)benzoic acid 95%
4-(Allylamino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD02252822
CAS Number: 53624-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR