2-Bromo-5-fluoro-3-methoxypyridine 98%

Supplier: Ambeed
Warning

A820739-100MG A820739-1G A820739-250MG A820739-5G
76993-434EA 128.37 USD
76993-434 76993-436 76993-438 76993-440
2-Bromo-5-fluoro-3-methoxypyridine 98%
2-Bromo-5-fluoro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MW: 206.01 g/mol
MDL Number: MFCD18257626
CAS Number: 1256806-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR