Methyl-5-hydroxy-2-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-hydroxy-2-methylbenzoate

A820179-100G A820179-1G A820179-250MG A820179-25G A820179-5G
77036-532EA 1723.97 USD
77036-532 77036-534 77036-536 77036-538 77036-540
Methyl-5-hydroxy-2-methylbenzoate 98%
Methyl-5-hydroxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD11226199
CAS Number: 73505-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR