2-Nitro-3-(trifluoromethyl)phenol ≥98%

Supplier: Ambeed
Warning

A819853-1G A819853-250MG
76993-430EA 41 USD
76993-430 76993-432
2-Nitro-3-(trifluoromethyl)phenol ≥98%
2-Nitro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₃
MDL Number: MFCD08282784
CAS Number: 386-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR