4-Chloro-6-(methoxymethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A818693-100MG A818693-1G A818693-250MG
76993-420EA 67.9 USD
76993-420 76993-422 76993-424
4-Chloro-6-(methoxymethyl)pyrimidine 97%
4-Chloro-6-(methoxymethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.58 g/mol
MDL Number: MFCD16707817
CAS Number: 3122-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR