8-Fluoroimidazo[1,2-a]pyridine 95%
Supplier: Ambeed
|
Warning
|
A817539-1G
A817539-250MG
77036-524EA
164.44
USD
77036-524
77036-526
8-Fluoroimidazo[1,2-a]pyridine 95%
8-Fluoroimidazo[1,2-a]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅FN₂ MW: 136.12 g/mol |
MDL Number:
MFCD09954954 CAS Number: 139022-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...