2-Bromo-4-iodophenol 96%

Supplier: Ambeed
Warning

A817327-100MG A817327-1G A817327-250MG A817327-25G A817327-5G
76993-410EA 35.01 USD
76993-410 76993-412 76993-414 76993-416 76993-418
2-Bromo-4-iodophenol 96%
2-Bromo-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
MW: 298.9038 g/mol
MDL Number: MFCD11870189
CAS Number: 133430-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR