5-Amino-3-bromo-2-chloropyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-6-chloropyridin-3-amine, 3-Bromo-2-chloro-5-pyridylamine
A816259-100G
A816259-10G
A816259-1G
A816259-25G
A816259-5G
77006-662EA
178.24
USD
77006-662
77006-664
77006-666
77006-668
77674-204
5-Amino-3-bromo-2-chloropyridine 98%
5-Amino-3-bromo-2-chloropyridine
Precaución: for Research Use Only
Formula:
C₅H₄BrClN₂ MW: 207.46 g/mol |
MDL Number:
MFCD03840433 CAS Number: 130284-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...