(4,5-Dichloro-1H-imidazol-2-yl)methanol 97%

Supplier: Ambeed
Warning

A815931-1G A815931-250MG
77019-490EA 45 USD
77019-490 77019-492
(4,5-Dichloro-1H-imidazol-2-yl)methanol 97%
(4,5-Dichloro-1H-imidazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₄Cl₂N₂O
MDL Number: MFCD18808554
CAS Number: 437658-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR