1,4-Di(4-pyridyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(1,4-Phenylene)bispyridine, 1,4-Di(pyridin-4-yl)benzene

A814723-100MG A814723-1G A814723-250MG A814723-5G
77019-482EA 12.74 USD
77019-482 77019-484 77019-486 77019-488
1,4-Di(4-pyridyl)benzene 97%
1,4-Di(4-pyridyl)benzene

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂
MW: 232.29 g/mol
MDL Number: MFCD00445845
CAS Number: 113682-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR