Benzoyl-DL-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Benzamido-3-phenylpropanoic acid, Bz-DL-Phe-OH

A814658-25G A814658-5G A814658-250MG
77036-508EA 161 USD
77036-508 77036-510 77873-275
Benzoyl-DL-phenylalanine 98%
Benzoyl-DL-phenylalanine

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₃
MDL Number: MFCD00037252
CAS Number: 2901-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR