Boc-(S)-3-amino-4-(4-trifluoromethylphenyl)butyric acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-((tert-Butoxycarbonyl)amino)-4-(4-(trifluoromethyl)phenyl)butanoic acid

A814404-100MG A814404-250MG
77036-504EA 50.93 USD
77036-504 77036-506
Boc-(S)-3-amino-4-(4-trifluoromethylphenyl)butyric acid ≥98%
Boc-(S)-3-amino-4-(4-trifluoromethylphenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₆H₂₀F₃NO₄
MW: 347.34 g/mol
MDL Number: MFCD01861070
CAS Number: 270065-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR