tert-Butyl 4-(3-ethoxy-3-oxopropyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(N-BOC-piperidin-4-yl)propioate

A814247-1G A814247-250MG A814247-5G
77036-498EA 35 USD
77036-498 77036-500 77036-502
tert-Butyl 4-(3-ethoxy-3-oxopropyl)piperidine-1-carboxylate 98%
tert-Butyl 4-(3-ethoxy-3-oxopropyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₇NO₄
MDL Number: MFCD08061335
CAS Number: 301232-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR