(2-Methylbutyl)boronic acid 95%

Supplier: Ambeed
Warning

A813965-100MG A813965-1G A813965-250MG
77036-492EA 56 USD
77036-492 77036-494 77036-496
(2-Methylbutyl)boronic acid 95%
(2-Methylbutyl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₁₃BO₂
MW: 115.96 g/mol
MDL Number: MFCD18861700
CAS Number: 444885-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR