3,3',3''-(1,3,5-Triazinane-1,3,5-triyl)tris(N,N-dimethylpropan-1-amine) 95%

Supplier: Ambeed
Danger

A813590-1G A813590-5G A813590-25G
77006-658EA 5 USD
77006-658 77006-660 77674-198
3,3',3''-(1,3,5-Triazinane-1,3,5-triyl)tris(N,N-dimethylpropan-1-amine) 95%
3,3',3''-(1,3,5-Triazinane-1,3,5-triyl)tris(N,N-dimethylpropan-1-amine)

Precaución: for Research Use Only

Formula: C₁₈H₄₂N₆
MDL Number: MFCD00059766
CAS Number: 15875-13-5
UN: 3267
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR