4-Amino-3-chloro-5-fluorobenzamide 95%

Supplier: Ambeed
Warning

A813471-100MG A813471-1G A813471-250MG
77019-468EA 70.02 USD
77019-468 77019-470 77019-472
4-Amino-3-chloro-5-fluorobenzamide 95%
4-Amino-3-chloro-5-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₆ClFN₂O
MW: 188.58 g/mol
MDL Number: MFCD27922987
CAS Number: 1823346-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR