1-Methylpiperazin-2-one hydrochloride 97%

Supplier: Ambeed
Warning

A812733-1G A812733-250MG A812733-5G
76993-392EA 42 USD
76993-392 76993-394 76993-396
1-Methylpiperazin-2-one hydrochloride 97%
1-Methylpiperazin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O
MDL Number: MFCD06662557
CAS Number: 109384-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR