6-(Methylthio)-1H-pyrazolo[3,4-d]pyrimidine 95%

Supplier: Ambeed
Warning

A811930-100MG A811930-250MG
77036-482EA 42.44 USD
77036-482 77036-484
6-(Methylthio)-1H-pyrazolo[3,4-d]pyrimidine 95%
6-(Methylthio)-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₆N₄S
MDL Number: MFCD11040464
CAS Number: 55084-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR