Benzyl 4-aminobenzoate 98%

Supplier: Ambeed
Warning

A811820-100MG A811820-250MG A811820-5G A811820-1G
76993-386EA 29 USD
76993-386 76993-388 76993-390 77595-292
Benzyl 4-aminobenzoate 98%
Benzyl 4-aminobenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MDL Number: MFCD00626894
CAS Number: 19008-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR