2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine hydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine Hydrochloride

A811361-1G A811361-250MG
76993-382EA 124 USD
76993-382 76993-384
2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine hydrochloride 96%
2,2,2-Trifluoro-1-(3-fluorophenyl)ethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₈ClF₄N
MW: 229.61 g/mol
MDL Number: MFCD10565658
CAS Number: 886368-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR