2,6-Bis(di-t-butylphosphinomethyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,6-Bis((di-tert-butylphosphino)methyl)pyridine
A811192-5G
A811192-250MG
76993-374EA
1050.3
USD
76993-374
77569-516
2,6-Bis(di-t-butylphosphinomethyl)pyridine 98%
2,6-Bis(di-t-butylphosphinomethyl)pyridine
Precaución: For research use only.
Formula:
C₂₃H₄₃NP₂ MW: 395.5418 g/mol |
MDL Number:
MFCD10565623 CAS Number: 338800-13-8 UN: 2813 ADR: 4.3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...