3-Amino-2-bromo-6-(trifluoromethyl)pyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-6-(trifluoromethyl)pyridin-3-amine, 5-Amino-6-bromo-α,α,α-trifluoro-2-picoline
A810094-1G
A810094-250MG
A810094-5G
A810094-25G
77036-456EA
15
USD
77036-456
77036-458
77036-460
77873-273
3-Amino-2-bromo-6-(trifluoromethyl)pyridine 97%
3-Amino-2-bromo-6-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₄BrF₃N₂ MW: 241.009 g/mol |
MDL Number:
MFCD11052334 CAS Number: 117519-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...