3-Bromo-2-fluoro-6-methoxypyridine 98%

Supplier: Ambeed
Warning

A809915-100G A809915-1G A809915-5G A809915-25G
77019-460EA 1067.27 USD
77019-460 77019-462 77573-590 77614-440
3-Bromo-2-fluoro-6-methoxypyridine 98%
3-Bromo-2-fluoro-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MW: 206.01 g/mol
MDL Number: MFCD16609956
CAS Number: 1227599-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR