5-Chlorosalicylic acid 98%

Supplier: Ambeed
Warning

Synonyms: CSA, 5-Chloro-2-hydroxybenzoic acid

A809692-1000G A809692-100G A809692-25G A809692-500G
77006-644EA 84.88 USD
77006-644 77006-646 77006-648 77006-650
5-Chlorosalicylic acid 98%
5-Chlorosalicylic acid

Precaución: for Research Use Only

Formula: C₇H₅ClO₃
MW: 172.57 g/mol
MDL Number: MFCD00002457
CAS Number: 321-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR