4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl)piperidine-3-carboxylate, 4-(3-Ethoxycarbonyl-1-piperidinylcarbonyl)benzeneboronic acid pinacol ester

A809664-1G A809664-250MG A809664-5G
76993-368EA 45.01 USD
76993-368 76993-370 76993-372
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester 98%
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₃₀BNO₅
MDL Number: MFCD06657751
CAS Number: 850411-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR