Boc-Orn(2-Cl-Z)-OH 98%

Supplier: Ambeed
Warning

A808866-1G A808866-25G A808866-5G
77036-442EA 17 USD
77036-442 77036-444 77036-446
Boc-Orn(2-Cl-Z)-OH 98%
Boc-Orn(2-Cl-Z)-OH

Precaución: for Research Use Only

Formula: C₁₈H₂₅ClN₂O₆
MDL Number: MFCD00076972
CAS Number: 118554-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR