1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A808356-250MG A808356-5G A808356-1G
77006-640EA 28.65 USD
77006-640 77006-642 77603-046
1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid 97%
1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅N₃O₄
MW: 171.111 g/mol
MDL Number: MFCD00222738
CAS Number: 92534-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR