4-(2-Phenylethylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Phenethylcarbamoyl)phenyl)boronic acid

A807546-1G A807546-250MG A807546-25G A807546-5G
76993-344EA 34 USD
76993-344 76993-346 76993-348 76993-350
4-(2-Phenylethylcarbamoyl)phenylboronic acid 98%
4-(2-Phenylethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆BNO₃
MDL Number: MFCD09027209
CAS Number: 330793-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR