4-(Pyridin-2-ylmethoxy)aniline 95%

Supplier: Ambeed
Warning

A806279-100MG A806279-1G A806279-250MG
77036-434EA 56.23 USD
77036-434 77036-436 77036-438
4-(Pyridin-2-ylmethoxy)aniline 95%
4-(Pyridin-2-ylmethoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MDL Number: MFCD04971040
CAS Number: 102137-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR