5-Fluorocytidine 95%

Supplier: Ambeed
Warning

A805463-10G A805463-1G A805463-5G A805463-25G
77006-616EA 48 USD
77006-616 77006-618 77006-620 77674-182
5-Fluorocytidine 95%
5-Fluorocytidine

Precaución: for Research Use Only

Formula: C₉H₁₂FN₃O₅
MDL Number: MFCD00210953
CAS Number: 2341-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR