tert-Butyl ((2R,3S)-2-(2,5-difluorophenyl)-5-oxotetrahydro-2H-pyran-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A804850-1G A804850-5G
76993-326EA 43.5 USD
76993-326 76993-328
tert-Butyl ((2R,3S)-2-(2,5-difluorophenyl)-5-oxotetrahydro-2H-pyran-3-yl)carbamate 95%
tert-Butyl ((2R,3S)-2-(2,5-difluorophenyl)-5-oxotetrahydro-2H-pyran-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₆H₁₉F₂NO₄
MW: 327.32 g/mol
MDL Number: MFCD20926096
CAS Number: 951127-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR