(1H-Inden-2-yl)boronic acid 95+%

Supplier: Ambeed
Warning

A804454-250MG A804454-5G A804454-1G
76993-322EA 78 USD
76993-322 76993-324 77891-554
(1H-Inden-2-yl)boronic acid 95+%
(1H-Inden-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BO₂
MDL Number: MFCD11506071
CAS Number: 312968-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR