3-Fluoro-2-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A804347-100MG A804347-1G A804347-250MG
76993-316EA 10 USD
76993-316 76993-318 76993-320
3-Fluoro-2-hydroxybenzonitrile 98%
3-Fluoro-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄FNO
MDL Number: MFCD12407120
CAS Number: 28177-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR