Amlodipine besylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylic acid 3-Ethyl 5-Methyl Ester Benzenesulphon, 3-Ethyl 5-Methyl 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate Benzenesulphonate, 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate benzenesulphonate
A804204-100G
A804204-1G
A804204-1MG
A804204-25G
A804204-5G
76993-308EA
110
USD
76993-308
76993-310
76993-312
76993-314
77886-791
Amlodipine besylate 97%
Amlodipine besylate
Precaución: for Research Use Only
Formula:
C₂₀H₂₅ClN₂O₅ |
MDL Number:
MFCD00887594 CAS Number: 111470-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...